N-(3-Acetylphenyl)butanamide

Supplier: Matrix Scientific

030043-500MG
101829-358EA 146.15 USD
101829-358
N-(3-Acetylphenyl)butanamide
N-(3-Acetylphenyl)butanamide

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD00448103
CAS Number: 96604-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR