1-(4-Cyclohexylphenyl)propan-1-one

Supplier: Matrix Scientific

030041-500MG
101829-354EA 116.92 USD
101829-354
1-(4-Cyclohexylphenyl)propan-1-one
1-(4-Cyclohexylphenyl)propan-1-one

Hazard: Irritant

Formula: C₁₅H₂₀O
Boiling Pt: 180…185 °C (3 Torr)
Melting Pt: 41 °C
MDL Number: MFCD02245138
CAS Number: 59721-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR