1-(4-Propylphenyl)propan-1-one

Supplier: Matrix Scientific

030033-500MG
101829-338EA 116.92 USD
101829-338
1-(4-Propylphenyl)propan-1-one
1-(4-Propylphenyl)propan-1-one

Hazard: Irritant

Formula: C₁₂H₁₆O
MDL Number: MFCD01993684
CAS Number: 56147-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR