4'-Ethylpropiophenone

Supplier: Matrix Scientific

Synonyms: 1-(4-Ethylphenyl)propan-1-one

030032-500MG
101829-336EA 116.92 USD
101829-336
4'-Ethylpropiophenone
4'-Ethylpropiophenone

Hazard: Irritant

Formula: C₁₁H₁₄O
MW: 162.2300 g/mol
MDL Number: MFCD00191646
CAS Number: 27465-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR