1-(3-Isopropoxyphenyl)ethanone

Supplier: Matrix Scientific

030030-500MG
101829-332EA 116.92 USD
101829-332
1-(3-Isopropoxyphenyl)ethanone
1-(3-Isopropoxyphenyl)ethanone

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01922015
CAS Number: 114590-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR