1-(3-Propoxyphenyl)ethanone

Supplier: Matrix Scientific

030029-500MG
101829-330EA 116.92 USD
101829-330
1-(3-Propoxyphenyl)ethanone
1-(3-Propoxyphenyl)ethanone

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01922000
CAS Number: 121704-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR