1-(3-Ethoxyphenyl)ethanone

Supplier: Matrix Scientific

030028-500MG
101829-328EA 116.92 USD
101829-328
1-(3-Ethoxyphenyl)ethanone
1-(3-Ethoxyphenyl)ethanone

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD01921953
CAS Number: 52600-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR