N-(3-Acetylphenyl)propanamide

Supplier: Matrix Scientific

030025-500MG
101829-322EA 146.15 USD
101829-322
N-(3-Acetylphenyl)propanamide
N-(3-Acetylphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD00791263
CAS Number: 39569-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR