N-(4-Acetylphenyl)propanamide

Supplier: Matrix Scientific

030024-500MG
101829-320EA 146.15 USD
101829-320
N-(4-Acetylphenyl)propanamide
N-(4-Acetylphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD00032404
CAS Number: 7470-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR