N-(3-Acetylphenyl)-2,2,3,3,4,4,4-heptafluorobutanamide

Supplier: Matrix Scientific

030023-500MG
101829-318EA 146.15 USD
101829-318
N-(3-Acetylphenyl)-2,2,3,3,4,4,4-heptafluorobutanamide
N-(3-Acetylphenyl)-2,2,3,3,4,4,4-heptafluorobutanamide

Hazard: Irritant

Formula: C₁₂H₈F₇NO₂
MDL Number: MFCD00783377
CAS Number: 333396-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR