N-(4-Acetylphenyl)nicotinamide

Supplier: Matrix Scientific

030020-500MG
101829-312EA 146.15 USD
101829-312
N-(4-Acetylphenyl)nicotinamide
N-(4-Acetylphenyl)nicotinamide

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂
MDL Number: MFCD00441250
CAS Number: 75075-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR