1-(4-Isobutoxyphenyl)ethanone

Supplier: Matrix Scientific

030015-500MG
101829-302EA 192.31 USD
101829-302
1-(4-Isobutoxyphenyl)ethanone
1-(4-Isobutoxyphenyl)ethanone

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD00995652
CAS Number: 24242-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR