N-(4-Acetylphenyl)cyclopropanecarboxamide

Supplier: Matrix Scientific

030011-500MG
101829-294EA 146.15 USD
101829-294
N-(4-Acetylphenyl)cyclopropanecarboxamide
N-(4-Acetylphenyl)cyclopropanecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD00245472
CAS Number: 23845-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR