N-(3-Acetylphenyl)-2,4-dichlorobenzamide

Supplier: Matrix Scientific

030005-500MG
101829-282EA 109.25 USD
101829-282
N-(3-Acetylphenyl)-2,4-dichlorobenzamide
N-(3-Acetylphenyl)-2,4-dichlorobenzamide

Hazard: Irritant

Formula: C₁₅H₁₁Cl₂NO₂
MDL Number: MFCD00422205
CAS Number: 316143-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR