N-(3-Acetylphenyl)-4-chlorobenzamide

Supplier: Matrix Scientific

030004-500MG
101829-280EA 146.15 USD
101829-280
N-(3-Acetylphenyl)-4-chlorobenzamide
N-(3-Acetylphenyl)-4-chlorobenzamide

Hazard: Irritant

Formula: C₁₅H₁₂ClNO₂
MDL Number: MFCD00791228
CAS Number: 223500-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR