N-(4-Acetylphenyl)-2,2,2-trifluoroacetamide
Supplier: Matrix Scientific
030002-500MG
101829-276EA
146.15
USD
101829-276
N-(4-Acetylphenyl)-2,2,2-trifluoroacetamide
N-(4-Acetylphenyl)-2,2,2-trifluoroacetamide
Hazard: Irritant
Formula:
C₁₀H₈F₃NO₂ |
MDL Number:
MFCD00780957 CAS Number: 24568-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...