N-(4-Acetylphenyl)-1-benzothiophene-3-carboxamide

Supplier: Matrix Scientific

029999-500MG
101829-270EA 193.85 USD
101829-270
N-(4-Acetylphenyl)-1-benzothiophene-3-carboxamide
N-(4-Acetylphenyl)-1-benzothiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₇H₁₃NO₂S
MDL Number: MFCD03419978
CAS Number: 886629-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR