N-(3-Acetylphenyl)-1-benzothiophene-3-carboxamide

Supplier: Matrix Scientific

029998-500MG
101829-268EA 144.9 USD
101829-268
N-(3-Acetylphenyl)-1-benzothiophene-3-carboxamide
N-(3-Acetylphenyl)-1-benzothiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₇H₁₃NO₂S
MDL Number: MFCD03419977
CAS Number: 886629-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR