N-(4-Acetylphenyl)-4,5-dimethylthiophene-3-carboxamide

Supplier: Matrix Scientific

029993-500MG
101829-258EA 193.85 USD
101829-258
N-(4-Acetylphenyl)-4,5-dimethylthiophene-3-carboxamide
N-(4-Acetylphenyl)-4,5-dimethylthiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₅H₁₅NO₂S
MDL Number: MFCD03419972
CAS Number: 895920-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR