N-(3-Acetylphenyl)-5-methylthiophene-3-carboxamide

Supplier: Matrix Scientific

029990-500MG
101829-252EA 144.9 USD
101829-252
N-(3-Acetylphenyl)-5-methylthiophene-3-carboxamide
N-(3-Acetylphenyl)-5-methylthiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₃NO₂S
MDL Number: MFCD03419969
CAS Number: 895920-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR