N-(3-Acetylphenyl)-1-methyl-1H-pyrazole-5-carboxamide

Supplier: Matrix Scientific

029989-500MG
101829-250EA 251.85 USD
101829-250
N-(3-Acetylphenyl)-1-methyl-1H-pyrazole-5-carboxamide
N-(3-Acetylphenyl)-1-methyl-1H-pyrazole-5-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
MDL Number: MFCD03419829
CAS Number: 522619-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR