N-(4-Acetylphenyl)-5-methylisoxazole-3-carboxamide

Supplier: Matrix Scientific

029987-500MG
101829-246EA 193.85 USD
101829-246
N-(4-Acetylphenyl)-5-methylisoxazole-3-carboxamide
N-(4-Acetylphenyl)-5-methylisoxazole-3-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₃
MDL Number: MFCD03233689
CAS Number: 438213-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR