1-Bicyclo[2.2.1]hept-2-ylethanone

Supplier: Matrix Scientific

029986-500MG
101829-244EA 109.25 USD
101829-244
1-Bicyclo[2.2.1]hept-2-ylethanone
1-Bicyclo[2.2.1]hept-2-ylethanone

Hazard: Irritant

Formula: C₉H₁₄O
MDL Number: MFCD00183741
CAS Number: 58654-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR