2-Acetyl-5-methylfuran

Supplier: Matrix Scientific

Synonyms: 1-(5-Methylfuran-2-yl)ethanone

029985-500MG
101829-242EA 109.25 USD
101829-242
2-Acetyl-5-methylfuran
2-Acetyl-5-methylfuran

Hazard: Irritant

Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00003243
CAS Number: 1193-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR