1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)ethanone

Supplier: Matrix Scientific

029982-500MG
101829-236EA 173.85 USD
101829-236
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)ethanone
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)ethanone

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD03167208
CAS Number: 946658-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR