1-(1-Ethyl-1H-pyrazol-4-yl)ethanone

Supplier: Matrix Scientific

029980-500MG
101829-232EA 193.85 USD
101829-232
1-(1-Ethyl-1H-pyrazol-4-yl)ethanone
1-(1-Ethyl-1H-pyrazol-4-yl)ethanone

Hazard: Irritant

Formula: C₇H₁₀N₂O
MDL Number: MFCD04967136
CAS Number: 925147-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR