4-Chloro-2,5-dimethylbenzenesulfonohydrazide

Supplier: Matrix Scientific

029977-500MG
101829-226EA 193.85 USD
101829-226
4-Chloro-2,5-dimethylbenzenesulfonohydrazide
4-Chloro-2,5-dimethylbenzenesulfonohydrazide

Hazard: Irritant

Formula: C₈H₁₁ClN₂O₂S
MDL Number: MFCD00117330
CAS Number: 219688-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR