2,4,5-Trichlorobenzenesulfonohydrazide

Supplier: Matrix Scientific

029976-500MG
101829-224EA 144.9 USD
101829-224
2,4,5-Trichlorobenzenesulfonohydrazide
2,4,5-Trichlorobenzenesulfonohydrazide

Hazard: Irritant

Formula: C₆H₅Cl₃N₂O₂S
MDL Number: MFCD00007584
CAS Number: 6655-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR