1-[(3,4-Dimethylphenyl)sulfonyl]piperidine-4-carbohydrazide

Supplier: Matrix Scientific

029969-500MG
101829-214EA 193.85 USD
101829-214
1-[(3,4-Dimethylphenyl)sulfonyl]piperidine-4-carbohydrazide
1-[(3,4-Dimethylphenyl)sulfonyl]piperidine-4-carbohydrazide

Hazard: Irritant

Formula: C₁₄H₂₁N₃O₃S
MDL Number: MFCD08097757
CAS Number: 898148-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR