1-[(2,5-Dichlorophenyl)sulfonyl]piperidine-4-carbohydrazide

Supplier: Matrix Scientific

029968-500MG
101829-212EA 193.85 USD
101829-212
1-[(2,5-Dichlorophenyl)sulfonyl]piperidine-4-carbohydrazide
1-[(2,5-Dichlorophenyl)sulfonyl]piperidine-4-carbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂N₃O₃S
MDL Number: MFCD02044341
CAS Number: 309278-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR