1-[(2,3,5,6-Tetramethylphenyl)sulfonyl]piperidine-4-carbohydrazide

Supplier: Matrix Scientific

029967-500MG
101829-210EA 193.85 USD
101829-210
1-[(2,3,5,6-Tetramethylphenyl)sulfonyl]piperidine-4-carbohydrazide
1-[(2,3,5,6-Tetramethylphenyl)sulfonyl]piperidine-4-carbohydrazide

Hazard: Irritant

Formula: C₁₆H₂₅N₃O₃S
MDL Number: MFCD09971928
CAS Number: 956576-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR