1-(2,3,4,5,6-Pentamethylphenylsulfonyl)piperidine-4-carbohydrazide

Supplier: Matrix Scientific

029966-500MG
101829-208EA 193.85 USD
101829-208
1-(2,3,4,5,6-Pentamethylphenylsulfonyl)piperidine-4-carbohydrazide
1-(2,3,4,5,6-Pentamethylphenylsulfonyl)piperidine-4-carbohydrazide

Hazard: Irritant

Formula: C₁₇H₂₇N₃O₃S
MDL Number: MFCD03946028
CAS Number: 590357-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR