1-[(4-Fluorophenyl)sulfonyl]piperidine-4-carbohydrazide

Supplier: Matrix Scientific

029961-500MG
101829-198EA 193.85 USD
101829-198
1-[(4-Fluorophenyl)sulfonyl]piperidine-4-carbohydrazide
1-[(4-Fluorophenyl)sulfonyl]piperidine-4-carbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₆FN₃O₃S
MDL Number: MFCD01821806
CAS Number: 312534-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR