1-((4-Chlorophenyl)sulfonyl)-4-pyridinecarbohydrazide
Supplier: Matrix Scientific
029960-500MG
101829-196EA
193.85
USD
101829-196
1-((4-Chlorophenyl)sulfonyl)-4-pyridinecarbohydrazide
1-((4-Chlorophenyl)sulphonyl)-4-pyridinecarbohydrazide
Hazard: Irritant
Formula:
C₁₂H₁₆ClN₃O₃S |
MDL Number:
MFCD00754333 CAS Number: 332400-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...