1-(Methylsulfonyl)piperidine-4-carbohydrazide

Supplier: Matrix Scientific

029958-500MG
101829-192EA 251.85 USD
101829-192
1-(Methylsulfonyl)piperidine-4-carbohydrazide
1-(Methylsulfonyl)piperidine-4-carbohydrazide

Hazard: Irritant

Formula: C₇H₁₅N₃O₃S
MDL Number: MFCD01572745
CAS Number: 312515-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR