2-[(2,4-Dichlorobenzyl)thio]acetohydrazide

Supplier: Matrix Scientific

029954-500MG
101829-184EA 109.25 USD
101829-184
2-[(2,4-Dichlorobenzyl)thio]acetohydrazide
2-[(2,4-Dichlorobenzyl)thio]acetohydrazide

Hazard: Irritant

Formula: C₉H₁₀Cl₂N₂OS
MDL Number: MFCD04058783
CAS Number: 669728-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR