2-[(4-Bromobenzyl)thio]acetohydrazide

Supplier: Matrix Scientific

029952-500MG
101829-180EA 146.15 USD
101829-180
2-[(4-Bromobenzyl)thio]acetohydrazide
2-[(4-Bromobenzyl)thio]acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁BrN₂OS
MDL Number: MFCD04058245
CAS Number: 669726-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR