2-[(2-Bromobenzyl)thio]acetohydrazide
Supplier: Matrix Scientific
029951-500MG
101829-178EA
146.15
USD
101829-178
2-[(2-Bromobenzyl)thio]acetohydrazide
2-[(2-Bromobenzyl)thio]acetohydrazide
Hazard: Irritant
Formula:
C₉H₁₁BrN₂OS |
MDL Number:
MFCD03946406 CAS Number: 590376-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...