2-[(4-Methylbenzyl)thio]acetohydrazide

Supplier: Matrix Scientific

029950-500MG
101829-176EA 109.25 USD
101829-176
2-[(4-Methylbenzyl)thio]acetohydrazide
2-[(4-Methylbenzyl)thio]acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂OS
MDL Number: MFCD04058429
CAS Number: 669726-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR