2-[(4-Fluorobenzyl)thio]acetohydrazide

Supplier: Matrix Scientific

029947-500MG
101829-170EA 146.15 USD
101829-170
2-[(4-Fluorobenzyl)thio]acetohydrazide
2-[(4-Fluorobenzyl)thio]acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁FN₂OS
MDL Number: MFCD04057603
CAS Number: 669709-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR