2-[(2-Chlorobenzyl)thio]acetohydrazide

Supplier: Matrix Scientific

029942-500MG
101829-160EA 146.15 USD
101829-160
2-[(2-Chlorobenzyl)thio]acetohydrazide
2-[(2-Chlorobenzyl)thio]acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁ClN₂OS
MDL Number: MFCD01103747
CAS Number: 401639-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR