5,6,7,8-Tetrahydro-4H-cyclohepta[b]thiophene-2-carbohydrazide

Supplier: Matrix Scientific

029941-500MG
101829-158EA 217.35 USD
101829-158
5,6,7,8-Tetrahydro-4H-cyclohepta[b]thiophene-2-carbohydrazide
5,6,7,8-Tetrahydro-4H-cyclohepta[b]thiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂OS
MDL Number: MFCD03944753
CAS Number: 588696-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR