6-Ethyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carbohydrazide

Supplier: Matrix Scientific

029939-500MG
101829-154EA 290.77 USD
101829-154
6-Ethyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carbohydrazide
6-Ethyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂OS
MDL Number: MFCD09971926
CAS Number: 956576-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR