4,5,6,7-Tetrahydro-1-benzothiophene-2-carbohydrazide

Supplier: Matrix Scientific

029937-500MG
101829-150EA 217.35 USD
101829-150
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbohydrazide
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₁₂N₂OS
MDL Number: MFCD04969092
CAS Number: 65361-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR