3-Chloro-4-methyl-1-benzothiophene-2-carbohydrazide

Supplier: Matrix Scientific

029933-500MG
101829-142EA 290.77 USD
101829-142
3-Chloro-4-methyl-1-benzothiophene-2-carbohydrazide
3-Chloro-4-methyl-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₉ClN₂OS
MDL Number: MFCD09971924
CAS Number: 956576-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR