4-Phenylsemicarbazide

Supplier: Matrix Scientific

Synonyms: N-PhenylhydrazinecarboxamidePhenylsemicarbazideAnilinoformylhydrazine

029931-500MG
101829-138EA 144.9 USD
101829-138
4-Phenylsemicarbazide
4-Phenylsemicarbazide

Hazard: Irritant

Formula: C₇H₉N₃O
MDL Number: MFCD00007590
CAS Number: 537-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR