Naphthalene-2-sulfonohydrazide

Supplier: Matrix Scientific

029927-500MG
101829-130EA 144.9 USD
101829-130
Naphthalene-2-sulfonohydrazide
Naphthalene-2-sulfonohydrazide

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂S
MDL Number: MFCD00596250
CAS Number: 10151-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR