3-Nitrobenzenesulfonohydrazide

Supplier: Matrix Scientific

029926-500MG
101829-128EA 251.85 USD
101829-128
3-Nitrobenzenesulfonohydrazide
3-Nitrobenzenesulfonohydrazide

Hazard: Irritant

Formula: C₆H₇N₃O₄S
MDL Number: MFCD01859908
CAS Number: 6655-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR