3,4-Dimethoxybenzenesulfonohydrazide

Supplier: Matrix Scientific

029925-500MG
101829-126EA 161.54 USD
101829-126
3,4-Dimethoxybenzenesulfonohydrazide
3,4-Dimethoxybenzenesulfonohydrazide

Hazard: Irritant

Formula: C₈H₁₂N₂O₄S
MDL Number: MFCD03423369
CAS Number: 23095-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR