2,4-Dimethoxybenzenesulfonohydrazide
Supplier: Matrix Scientific
029923-500MG
101829-122EA
155.38
USD
101829-122
2,4-Dimethoxybenzenesulfonohydrazide
2,4-Dimethoxybenzenesulfonohydrazide
Hazard: Irritant
Formula:
C₈H₁₂N₂O₄S |
MDL Number:
MFCD03423367 CAS Number: 73737-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...